siRNA Details
| Gene Symbol: Dsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023842.2 | NP_076331.2 | 24 | 7910 | ||
| GenBank mRNA | AK077574.1 | - | 1 | 412 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dsp| Gene Symbol: Dsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023842.2 | NP_076331.2 | 24 | 7910 | ||
| GenBank mRNA | AK077574.1 | - | 1 | 412 | ||