siRNA Details
| Gene Symbol: Upk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026815.2 | NP_081091.1 | 3, 4 | 310 | ||
| GenBank mRNA | AF262335.1 | - | 2, 3 | 282 | ||
| AK004014.1 | - | 3, 4 | 311 | |||
| BC132047.1 | - | 2, 3 | 302 | 40130324 | ||
| BC138390.1 | - | 2, 3 | 302 | 8861410 | ||

Upk1a
Genomic Map