siRNA Details
| Gene Symbol: Upk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026815.2 | NP_081091.1 | 8 | 770 | ||
| GenBank mRNA | AF262335.1 | - | 7 | 742 | ||
| AK004014.1 | - | 8 | 771 | |||
| BC132047.1 | - | 7 | 762 | 40130324 | ||
| BC138390.1 | - | 7 | 762 | 8861410 | ||

Upk1a
Genomic Map