siRNA Details
| Gene Symbol: Arrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177231.2 | NP_796205.1 | 15, 16 | 1441 | ||
| NM_178220.3 | NP_835738.1 | 14, 15 | 1417 | |||
| GenBank mRNA | AK036421.1 | - | 4, 5 | 348 | ||
| AK052588.1 | - | 15, 16 | 1218 | |||
| AK090090.1 | - | 14, 15 | 1244 | |||
| AK144054.1 | - | 15, 16 | 1240 | |||
| AK147508.1 | - | 15, 16 | 1442 | |||
| AK147566.1 | - | 15, 16 | 1463 | |||
| AK156213.1 | - | 4, 5 | 351 | |||
| AK165514.1 | - | 15, 16 | 1240 | |||
| BC108969.1 | - | 14, 15 | 1184 | 40047224 | ||
| BC108970.1 | - | 14, 15 | 1184 | 40047225 | ||

Arrb1
Genomic Map