siRNA Details
| Gene Symbol: Itga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033228.3 | NP_001028400.2 | 2 | 550 | ||
| GenBank mRNA | AK029105.1 | - | 2 | 466 | ||
| AK035870.1 | - | 2 | 551 | |||
| AK043769.1 | - | 2 | 489 | |||
| AK137169.1 | - | 2 | 316 | |||
| AK141204.1 | - | 2 | 518 | |||
| AK165357.1 | - | 2 | 297 | |||
| AK169964.1 | - | 2 | 465 | |||
| BC014765.1 | - | 2 | 414 | |||

Itga1
Genomic Map