siRNA Details
| Gene Symbol: Pgm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028352.4 | NP_082628.3 | 10, 11 | 1364 | ||
| GenBank mRNA | AK013402.1 | - | 10, 11 | 1347 | ||
| AK028066.1 | - | 10, 11 | 1354 | |||
| AK032776.1 | - | 10, 11 | 1316 | |||
| AK036561.1 | - | 10, 11 | 1350 | |||
| AK043547.1 | - | 10, 11 | 1365 | |||
| AK049337.1 | - | 10, 11 | 1316 | |||
| AK051706.1 | - | 10, 11 | 1304 | |||
| AK153501.1 | - | 9, 10 | 1184 | |||
| AK160913.1 | - | 10, 11 | 1365 | |||
| AK165513.1 | - | 10, 11 | 1349 | |||
| AK169082.1 | - | 10, 11 | 1329 | |||
| AK169141.1 | - | 10, 11 | 1323 | |||
| AK169787.1 | - | 10, 11 | 1338 | |||
| BC138700.1 | - | 10, 11 | 1289 | 8861722 | ||

Pgm3
Genomic Map