siRNA Details
| Gene Symbol: Pgm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163746.1 | NP_001157218.1 | 5 | 612 | ||
| NM_028352.4 | NP_082628.3 | 5 | 612 | |||
| GenBank mRNA | AK013402.1 | - | 5 | 595 | ||
| AK028066.1 | - | 5 | 602 | |||
| AK032776.1 | - | 5 | 564 | |||
| AK036561.1 | - | 5 | 598 | |||
| AK043547.1 | - | 5 | 613 | |||
| AK049337.1 | - | 5 | 564 | |||
| AK049758.1 | - | 5 | 599 | |||
| AK051706.1 | - | 5 | 552 | |||
| AK153501.1 | - | 4 | 432 | |||
| AK160913.1 | - | 5 | 613 | |||
| AK165513.1 | - | 5 | 597 | |||
| AK169082.1 | - | 5 | 577 | |||
| AK169141.1 | - | 5 | 571 | |||
| AK169787.1 | - | 5 | 586 | |||
| BC138700.1 | - | 5 | 537 | 8861722 | ||

Pgm3
Genomic Map