siRNA Details
| Gene Symbol: Asl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133768.4 | NP_598529.1 | 14 | 1121 | ||
| GenBank mRNA | AK010922.1 | - | 14 | 1133 | ||
| AK076453.1 | - | 14 | 1136 | |||
| AK138423.1 | - | 14 | 1118 | |||
| AK140160.1 | - | 14 | 1136 | |||
| AK148764.1 | - | 14 | 1105 | |||
| AK168739.1 | - | 14 | 1116 | |||
| AK186697.1 | - | 2 | 117 | |||
| AK215777.1 | - | 2 | 117 | |||
| BC016670.1 | - | 14 | 1121 | 2648753 | ||

Asl
Genomic Map