siRNA Details
| Gene Symbol: Abr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198018.2 | NP_932135.1 | 2 | 581 | ||
| NM_198894.2 | NP_942597.1 | 3 | 467 | |||
| NM_198895.2 | NP_942598.1 | 3 | 588 | |||
| GenBank mRNA | AK044797.1 | - | 2 | 1381 | ||
| AK052216.1 | - | 2 | 799 | |||
| AK142364.1 | - | 3 | 570 | |||
| AK153615.1 | - | 2 | 1249 | |||
| BC058708.1 | - | 2 | 581 | 6839300 | ||
| CF535448.1 | - | 3 | 456 | |||
| CF536236.1 | - | 3 | 578 | |||

Abr
Genomic Map