siRNA Details
| Gene Symbol: Tpgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148934.2 | NP_683736.1 | 2 | 733 | ||
| GenBank mRNA | AF303106.1 | - | 2 | 733 | ||
| AK153467.1 | - | 2 | 660 | |||
| BC138515.1 | - | 2 | 774 | 8861535 | ||
| BC138516.1 | - | 2 | 774 | 8861536 | ||

Tpgs1
Genomic Map