siRNA Details
| Gene Symbol: Dek | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025900.2 | NP_080176.2 | 6 | 654 | ||
| GenBank mRNA | AK007546.1 | - | 3 | 203 | ||
| AK012615.1 | - | 6 | 595 | |||
| AK012877.1 | - | 6 | 589 | |||
| AK033451.1 | - | 6 | 544 | |||
| AK154013.1 | - | 6 | 636 | |||
| BC048844.1 | - | 6 | 604 | |||
| BC055451.1 | - | 6 | 600 | 4164972 | ||

Dek
Genomic Map