siRNA Details
| Gene Symbol: Phka2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177878.1 | NP_001171349.1 | 11 | 1457 | ||
| NM_001177879.1 | NP_001171350.1 | 12 | 1271 | |||
| NM_172783.3 | NP_766371.1 | 11 | 1457 | |||
| GenBank mRNA | AK052346.1 | - | 11 | 1202 | ||
| AK052992.1 | - | 11 | 1458 | |||
| AK165506.1 | - | 12 | 1272 | |||
| BC050040.1 | - | 12 | 1230 | 6431743 | ||
| BC051167.1 | - | 6 | 697 | |||
| BC058254.1 | - | 11 | 1426 | |||

Phka2
Genomic Map