siRNA Details
| Gene Symbol: Scgb2b26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178308.2 | NP_840093.1 | 2 | 112 | ||
| GenBank mRNA | AY293282.1 | - | 1 | 22 | ||
| BC035222.1 | - | 2 | 98 | 4192901 | ||
| BC107292.1 | - | 2 | 100 | 40053055 | ||
| BC107293.1 | - | 2 | 100 | 40053056 | ||
| BC111446.1 | - | 2 | 112 | 40053210 | ||
| BC134362.1 | - | 2 | 118 | 40134659 | ||
| BC148302.2 | - | 2 | 118 | 40134662 | ||

Scgb2b26
Genomic Map