siRNA Details
| Gene Symbol: Glra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080438.2 | NP_536686.2 | 10 | 1384 | ||
| GenBank mRNA | AY230204.1 | - | 10 | 1323 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glra3| Gene Symbol: Glra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080438.2 | NP_536686.2 | 10 | 1384 | ||
| GenBank mRNA | AY230204.1 | - | 10 | 1323 | ||