siRNA Details
| Gene Symbol: Slc9a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177084.3 | NP_796058.1 | 5 | 1646 | ||
| GenBank mRNA | AF139195.1 | - | 3 | 448 | ||
| AK085681.1 | - | 5 | 1672 | |||
| AK141534.1 | - | 5 | 1647 | |||
| AK170522.1 | - | 3 | 485 | |||
| BC120543.1 | - | 5 | 1564 | 40129466 | ||

Slc9a4
Genomic Map