siRNA Details
| Gene Symbol: Hlcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139145.4 | NP_631884.1 | 7 | 1784 | ||
| GenBank mRNA | AB066227.1 | - | 6 | 1644 | ||
| AK052424.1 | - | 8 | 2080 | |||
| AK158212.1 | - | 8 | 1901 | |||
| AK168925.1 | - | 4 | 679 | |||
| BC050090.1 | - | 7 | 1785 | 30012868 | ||

Hlcs
Genomic Map