siRNA Details
| Gene Symbol: M1ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033079.2 | NP_149070.2 | 5 | 768 | ||
| GenBank mRNA | AF084364.1 | - | 4 | 737 | ||
| AK131954.1 | - | 5 | 768 | |||
| BC119803.1 | - | 4 | 695 | 40107692 | ||
| BC122521.1 | - | 4 | 700 | 40107688 | ||

M1ap
Genomic Map