siRNA Details
| Gene Symbol: Mbd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033072.2 | NP_149063.2 | 4 | 309 | ||
| GenBank mRNA | AK158797.1 | - | 4 | 433 | ||
| BC059015.1 | - | 4 | 309 | |||
| BC096027.1 | - | 1 | 51 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mbd6| Gene Symbol: Mbd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033072.2 | NP_149063.2 | 4 | 309 | ||
| GenBank mRNA | AK158797.1 | - | 4 | 433 | ||
| BC059015.1 | - | 4 | 309 | |||
| BC096027.1 | - | 1 | 51 | |||