siRNA Details
| Gene Symbol: Abca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013454.3 | NP_038482.3 | 4 | 598 | ||
| GenBank mRNA | AK051920.1 | - | 4 | 599 | ||
| X75926.1 | - | 3 | 361 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abca1| Gene Symbol: Abca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013454.3 | NP_038482.3 | 4 | 598 | ||
| GenBank mRNA | AK051920.1 | - | 4 | 599 | ||
| X75926.1 | - | 3 | 361 | |||