siRNA Details
| Gene Symbol: Acadm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007382.5 | NP_031408.1 | 5 | 525 | ||
| GenBank mRNA | AK131793.1 | - | 5 | 327 | ||
| AK146444.1 | - | 5 | 330 | |||
| BC013498.1 | - | 5 | 310 | 4222450 | ||
| CX232934.1 | - | 5 | 516 | |||
| U07159.1 | - | 5 | 314 | |||

Acadm
Genomic Map