siRNA Details
Gene Symbol: Vmn1r52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053222.1 | NP_444452.1 | 1 | 862 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn1r52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053222.1 | NP_444452.1 | 1 | 862 |