siRNA Details
| Gene Symbol: Vmn1r53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053226.2 | NP_444456.2 | 1 | 908 | ||
| GenBank mRNA | BC125605.1 | - | 1 | 908 | 40130120 | |
| BC132002.1 | - | 1 | 908 | 40130279 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r53| Gene Symbol: Vmn1r53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053226.2 | NP_444456.2 | 1 | 908 | ||
| GenBank mRNA | BC125605.1 | - | 1 | 908 | 40130120 | |
| BC132002.1 | - | 1 | 908 | 40130279 | ||