siRNA Details
| Gene Symbol: Vmn1r44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053227.2 | NP_444457.2 | 1 | 298 | ||
| GenBank mRNA | BC107184.1 | - | 1 | 298 | ||
| BC107185.1 | - | 1 | 298 | |||
| BC151088.1 | - | 1 | 395 | 9088001 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r44| Gene Symbol: Vmn1r44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053227.2 | NP_444457.2 | 1 | 298 | ||
| GenBank mRNA | BC107184.1 | - | 1 | 298 | ||
| BC107185.1 | - | 1 | 298 | |||
| BC151088.1 | - | 1 | 395 | 9088001 | ||