siRNA Details
Gene Symbol: Slc33a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272035.1 | NP_001258964.1 | 2 | 1365 | ||
NM_015728.5 | NP_056543.2 | 2 | 1365 | |||
GenBank mRNA | AK144703.1 | - | 2 | 1231 | ||
BC005616.1 | - | 2 | 942 | 3154886 | ||
BC054395.1 | - | 2 | 1365 | 6492236 | ||
BC063104.1 | - | 1298 | 5042025 |
