siRNA Details
| Gene Symbol: Acox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271898.1 | NP_001258827.1 | 1 | 383 | ||
| NM_015729.3 | NP_056544.2 | 1 | 383 | |||
| GenBank mRNA | AB034914.1 | - | 1 | 72 | ||
| AF006688.1 | - | 1 | 122 | |||
| AK040566.1 | - | 1 | 86 | |||
| AK051539.1 | - | 1 | 56 | |||
| AK054446.1 | - | 1 | 159 | |||
| AK128941.1 | - | 1 | 85 | |||
| BC054727.1 | - | 1 | 51 | |||
| BC056448.1 | - | 1 | 383 | 5704873 | ||

Acox1
Genomic Map