siRNA Details
| Gene Symbol: Zbtb21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081684.1 | NP_001075153.1 | 2 | 226 | ||
| NM_001081685.1 | NP_001075154.1 | 3 | 369 | |||
| NM_175428.3 | NP_780637.2 | 3 | 369 | |||
| GenBank mRNA | AK031118.1 | - | 3 | 358 | ||
| AK034057.1 | - | 3 | 370 | |||
| AK129313.1 | - | 2 | 203 | |||
| AK145072.1 | - | 2 | 221 | |||
| BC027135.1 | - | 1 | 23 | 5360683 | ||
| BC051176.1 | - | 2 | 218 | |||

Zbtb21
Genomic Map