siRNA Details
Gene Symbol: Crip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053250.2 | NP_444480.1 | 4 | 220 | ||
NM_181664.2 | NP_858050.1 | 4 | 220 | |||
GenBank mRNA | AF367970.1 | - | 4 | 220 | ||
AF367971.1 | - | 4 | 220 | |||
AK080272.1 | - | 4 | 220 | |||
AY555745.1 | - | 4 | 211 | |||
BC116360.1 | - | 4 | 220 | 40099973 |
