siRNA Details
| Gene Symbol: Ehbp1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114595.1 | NP_001108067.1 | 6 | 859 | ||
| NM_001114596.1 | NP_001108068.1 | 6 | 859 | |||
| NM_001114597.1 | NP_001108069.1 | 6 | 859 | |||
| NM_053252.3 | NP_444482.2 | 6 | 859 | |||
| GenBank mRNA | AF305087.1 | - | 6 | 812 | ||
| AF305088.1 | - | 6 | 812 | |||
| AF305089.1 | - | 6 | 812 | |||
| AF305090.1 | - | 6 | 812 | |||
| AK087817.1 | - | 2 | 101 | |||
| AK089917.1 | - | 6 | 820 | |||
| AK169974.1 | - | 6 | 825 | |||
| AK194109.1 | - | 1 | 20 | |||
| BC004660.1 | - | 6 | 808 | 3497913 | ||

Ehbp1l1
Genomic Map