siRNA Details
| Gene Symbol: Acvr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007396.4 | NP_031422.3 | 9 | 1776 | ||
| GenBank mRNA | AK041246.1 | - | 9 | 1729 | ||
| AK143186.1 | - | 10 | 1875 | |||
| BC138823.1 | - | 9 | 1437 | 8861845 | ||
| M65287.1 | - | 9 | 1183 | |||

Acvr2a
Genomic Map