siRNA Details
| Gene Symbol: Adam4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009620.1 | NP_033750.1 | 1 | 1248 | ||
| GenBank mRNA | AK076647.1 | - | 1 | 1309 | ||
| AY158688.1 | - | 1 | 1248 | |||
| BC150159.1 | - | 1 | 1252 | 40141514 | ||
| BC150481.1 | - | 1 | 1252 | 40141515 | ||
| U22058.1 | - | 1 | 345 | |||

Adam4
Genomic Map