siRNA Details
| Gene Symbol: Adam7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007402.2 | NP_031428.2 | 14 | 1498 | ||
| GenBank mRNA | AK136692.1 | - | 14 | 1497 | ||
| KF144369.1 | - | 2 | 221 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam7| Gene Symbol: Adam7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007402.2 | NP_031428.2 | 14 | 1498 | ||
| GenBank mRNA | AK136692.1 | - | 14 | 1497 | ||
| KF144369.1 | - | 2 | 221 | |||