siRNA Details
Gene Symbol: Adra2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009633.3 | NP_033763.2 | 1 | 1382 | ||
GenBank mRNA | AF332049.1 | - | 1 | 1088 | ||
AF332050.1 | - | 1 | 1088 | |||
BC066862.1 | - | 1 | 1382 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
