siRNA Details
| Gene Symbol: Crisp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009638.3 | NP_033768.3 | 8 | 728 | ||
| GenBank mRNA | AK009449.1 | - | 8 | 729 | ||
| BC011150.1 | - | 8 | 711 | 4165956 | ||
| L05559.1 | - | 8 | 698 | |||
| M92849.1 | - | 8 | 694 | |||

Crisp1
Genomic Map