siRNA Details
| Gene Symbol: Crisp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009639.2 | NP_033769.1 | 581 | |||
| GenBank mRNA | BC022573.1 | - | 7 | 602 | 4925349 | |
| BC114345.2 | - | 583 | ||||
| BC132536.1 | - | 580 | 40130813 | |||
| BC132538.1 | - | 580 | 40130815 | |||
| L05560.1 | - | 581 | ||||
| M92850.1 | - | 7 | 589 | |||

Crisp3
Genomic Map