siRNA Details
| Gene Symbol: Akap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035532.2 | NP_001030609.1 | 2 | 1407 | ||
| NM_001035533.2 | NP_001030610.1 | 1 | 1348 | |||
| NM_009649.3 | NP_033779.2 | 2 | 1407 | |||
| GenBank mRNA | AF033274.1 | - | 1 | 1750 | ||
| AF033275.1 | - | 1 | 1750 | |||
| AF033276.1 | - | 1 | 1750 | |||
| AK041873.1 | - | 2 | 1397 | |||
| AK147420.1 | - | 2 | 1375 | |||
| AK173063.1 | - | 1 | 1868 | |||
| Gene Symbol: Pakap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077020.1 | - | 3 | 1812 | ||

Akap2
Genomic Map