siRNA Details
| Gene Symbol: Akap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042542.2 | NP_001036007.1 | 5 | 684 | ||
| NM_009651.4 | NP_033781.2 | 5 | 680 | |||
| GenBank mRNA | BC130271.1 | - | 5 | 612 | 40143193 | |
| U07423.1 | - | 5 | 653 | |||
| U10341.1 | - | 5 | 685 | |||

Akap4
Genomic Map