siRNA Details
| Gene Symbol: Abcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007435.1 | NP_031461.1 | 6 | 2028 | ||
| GenBank mRNA | AK088792.1 | - | 6 | 1898 | ||
| BC011273.1 | - | 6 | 1847 | 3710503 | ||
| BC079840.1 | - | 6 | 1906 | 6810147 | ||
| Z33637.1 | - | 6 | 2028 | |||

Abcd1
Genomic Map