siRNA Details
| Gene Symbol: Aldh1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013467.3 | NP_038495.2 | 4 | 360 | ||
| GenBank mRNA | BC035322.1 | - | 4 | 354 | ||
| BC044729.2 | - | 4 | 361 | |||
| BC054386.1 | - | 4 | 360 | 6477503 | ||
| M74570.1 | - | 4 | 374 | |||
| S75713.1 | - | 4 | 330 | |||

Aldh1a1
Genomic Map