siRNA Details
| Gene Symbol: Akr1c20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054080.1 | NP_473421.1 | 1 | 86 | ||
| GenBank mRNA | AB097220.1 | - | 1 | 74 | ||
| AK012162.1 | - | 1 | 86 | |||
| BC021607.1 | - | 1 | 82 | 5098938 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akr1c20| Gene Symbol: Akr1c20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054080.1 | NP_473421.1 | 1 | 86 | ||
| GenBank mRNA | AB097220.1 | - | 1 | 74 | ||
| AK012162.1 | - | 1 | 86 | |||
| BC021607.1 | - | 1 | 82 | 5098938 | ||