siRNA Details
| Gene Symbol: Alox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009662.2 | NP_033792.1 | 2 | 442 | ||
| GenBank mRNA | AK089344.1 | - | 2 | 396 | ||
| AK136868.1 | - | 2 | 410 | |||
| AK137481.1 | - | 2 | 400 | |||
| AK169938.1 | - | 2 | 399 | |||
| AK170016.1 | - | 2 | 299 | |||
| AK170434.1 | - | 2 | 399 | |||
| AK170879.1 | - | 2 | 398 | |||
| AK171191.1 | - | 2 | 391 | |||
| AK171413.1 | - | 2 | 399 | |||
| BC139102.1 | - | 2 | 369 | 8862124 | ||
| BC141213.1 | - | 2 | 369 | 9055825 | ||
| L42198.1 | - | 2 | 288 | |||

Alox5
Genomic Map