siRNA Details
| Gene Symbol: Alpk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054085.2 | NP_473426.2 | 8, 9 | 4058 | ||
| GenBank mRNA | AF338872.1 | - | 8, 9 | 4052 | ||
| AK220384.1 | - | 4, 5 | 2534 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alpk3| Gene Symbol: Alpk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054085.2 | NP_473426.2 | 8, 9 | 4058 | ||
| GenBank mRNA | AF338872.1 | - | 8, 9 | 4052 | ||
| AK220384.1 | - | 4, 5 | 2534 | |||