siRNA Details
| Gene Symbol: Ambn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017250.1 | - | 11 | 903 | ||
| BC087927.1 | - | 11 | 947 | 30504437 | ||
| U65021.1 | - | 11 | 882 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ambn| Gene Symbol: Ambn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017250.1 | - | 11 | 903 | ||
| BC087927.1 | - | 11 | 947 | 30504437 | ||
| U65021.1 | - | 11 | 882 | |||