siRNA Details
| Gene Symbol: Cts3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026906.3 | NP_081182.2 | 7 | 1045 | ||
| NM_054092.2 | NP_473433.2 | 7 | 970 | |||
| GenBank mRNA | AF456464.1 | - | 6 | 830 | ||
| AK005389.1 | - | 7 | 968 | |||
| AY034574.1 | - | 7 | 984 | |||
| BC099388.1 | - | 7 | 939 | 30786828 | ||

Cts3
Genomic Map