siRNA Details
| Gene Symbol: Ttyh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053273.2 | NP_444503.2 | 2 | 244 | ||
| GenBank mRNA | AF329682.1 | - | 2 | 187 | ||
| AK152172.1 | - | 2 | 196 | |||
| AK154835.1 | - | 2 | 252 | |||
| AK155166.1 | - | 2 | 245 | |||
| AK156393.1 | - | 2 | 197 | |||
| AK168408.1 | - | 2 | 197 | |||
| BC060740.1 | - | 2 | 187 | 4158813 | ||
| BY352523.1 | - | 2 | 282 | |||
| BY740853.1 | - | 2 | 224 | |||

Ttyh2
Genomic Map