siRNA Details
| Gene Symbol: Mat1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133653.3 | NP_598414.1 | 8 | 1260 | ||
| GenBank mRNA | AK149542.1 | - | 7 | 1011 | ||
| AK153832.1 | - | 7 | 1014 | |||
| BC011211.1 | - | 7 | 998 | 4196928 | ||
| L13622.1 | - | 7 | 1012 | |||

Mat1a
Genomic Map