siRNA Details
| Gene Symbol: Ank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110783.3 | NP_001104253.1 | 38 | 4851 | ||
| NM_001277280.2 | NP_001264209.1 | 37 | 4827 | |||
| NM_001277281.2 | NP_001264210.1 | 37 | 4548 | |||
| NM_001277284.2 | NP_001264213.1 | 37 | 4548 | |||
| NM_001277286.2 | NP_001264215.1 | 38 | 4837 | |||
| NM_001277289.2 | NP_001264218.1 | 38 | 4837 | |||
| NM_031158.4 | NP_112435.2 | 38 | 4837 | |||
| GenBank mRNA | AK078124.1 | - | 2 | 1127 | ||
| AK147278.1 | - | 38 | 4806 | |||
| AK147483.1 | - | 38 | 4833 | |||
| AK147597.1 | - | 38 | 4785 | |||
| AK158131.1 | - | 37 | 4820 | |||
| BC079910.1 | - | 37 | 4519 | 30664024 | ||
| BC138029.1 | - | 38 | 4571 | 8861048 | ||
| BC145102.1 | - | 38 | 4473 | |||
| BC171944.1 | - | 38 | 4571 | 9054230 | ||
| M84756.1 | - | 37 | 4525 | |||
| X69063.1 | - | 39 | 4821 | |||

Ank1
Genomic Map