siRNA Details
| Gene Symbol: Ap2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077264.1 | NP_001070732.1 | 19 | 2583 | ||
| NM_007458.2 | NP_031484.1 | 20 | 2649 | |||
| GenBank mRNA | AK135424.1 | - | 19 | 2512 | ||
| AK155515.1 | - | 19 | 2520 | |||
| BC031433.1 | - | 20 | 2599 | 4481643 | ||
| Gene Symbol: Fuz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035030.1 | - | 1340 | |||
| AK045745.1 | - | 1570 | ||||

Ap2a1
Genomic Map