siRNA Details
| Gene Symbol: Ap3s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009682.3 | NP_033812.3 | 2 | 198 | ||
| GenBank mRNA | AK028372.1 | - | 2 | 181 | ||
| AK037336.1 | - | 2 | 176 | |||
| AK083911.1 | - | 2 | 187 | |||
| AK150085.1 | - | 2 | 191 | |||
| BC060236.1 | - | 2 | 159 | 6853219 | ||
| BY127127.1 | - | 2 | 201 | |||
| U91933.1 | - | 2 | 153 | |||

Ap3s2
Genomic Map