siRNA Details
| Gene Symbol: Speg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085370.1 | NP_001078839.1 | 3 | 1469 | ||
| NM_001085371.1 | NP_001078840.1 | 6 | 1936 | |||
| NM_007463.4 | NP_031489.4 | 5 | 2369 | |||
| GenBank mRNA | AF215896.1 | - | 5 | 2369 | ||
| AK147475.1 | - | 3 | 1474 | |||
| AK164360.1 | - | 6 | 1915 | |||
| BC062643.1 | - | 1 | 61 | |||

Speg
Genomic Map