siRNA Details
| Gene Symbol: Speg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085370.1 | NP_001078839.1 | 7 | 2043 | ||
| NM_001085371.1 | NP_001078840.1 | 10 | 2510 | |||
| NM_001173477.1 | NP_001166948.1 | 4 | 523 | |||
| NM_007463.4 | NP_031489.4 | 9 | 2943 | |||
| GenBank mRNA | AF215896.1 | - | 9 | 2943 | ||
| AK147475.1 | - | 7 | 2048 | |||
| AK164360.1 | - | 10 | 2489 | |||
| BC048698.1 | - | 4 | 420 | |||
| BC062643.1 | - | 5 | 635 | |||
| CK622939.1 | - | 4 | 470 | |||
| U57098.1 | - | 3 | 350 | |||

Speg
Genomic Map